Geneclean Glassmilk
Geneclean Glassmilk
Actual product may vary
MP Biomedicals
Geneclean Glassmilk
GeneClean Glassmilk
Have Questions?
Catalog number MPB111001204
Price (SGD)
318.00
EA
Add to cart
Price (SGD)
318.00
EA
Add to cart
  • Glassmilk slurry allows flexibility of reaction size and volume.
  • GeneClean Glassmilk is a specially prepared aqueous suspension of proprietary silica matrix.
  • GeneClean Glassmilk is supplied with GeneClean Kit, which will bind to radioactive isotope-labeled ssDNA or dsDNA that is purified from agarose thus resulting in purified DNA for further use.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Product TypeKits and Assays
DescriptionGeneclean™glassmilk™ is a specially prepared aqueous suspension of proprietary silica matrix.
Packaging Quantity1.25 mL
Unit SizeEA