Mycoplasma Removal Agent (MRA)
Mycoplasma Removal Agent (MRA)
Actual product may vary
MP Biomedicals
Mycoplasma Removal Agent (MRA)
Mycoplasma Removal Agent (MRA) is a derivative of the quinoline family of antibiotics and eliminates mycoplasma contamination in cell cultures by inhibiting mycoplasma DNA gyrase.
Have Questions?
Catalog NumberQuantity
MPB0930500445 mL
Catalog number MPB093050044
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
  • Derivative of the quinoline family of antibiotics and eliminates mycoplasma infection by inhibiting mycoplasma DNA gyrase
  • Supplied as a ready-to-use solution
  • Cited by the European Collection of Animal Cell Cultures (Porton Down, U.K.) as being superior to all other current products for the eradication of mycoplasma
  • Cell toxicity is low following treatment at the recommended concentration of 0.5 μg/mL
  • Eliminates multiple mycoplasma species within one week using the lowest dosage
  • Prevents recontamination of the culture at 0.1 μg/mL
  • Maintains cell viability
  • Compatible with most mammalian cell lines
  • Sterility and low cytotoxicity
Specifications
Concentration50 µg/mL
FormClear, light yellow solution
Product TypeMycoplasma Removal
SterilitySterile
For Use With (Application)Mycoplasma elimination
Quantity5 mL
Unit SizeEA
Contents & Storage
Store at Room Temperature (15 to 30°C). Protect from light.