Serum, Type AB, Human (HCV, HIV, HbsAG Free)
Serum, Type AB, Human (HCV, HIV, HbsAG Free)
Actual product may vary
MP Biomedicals
Serum, Type AB, Human (HCV, HIV, HbsAG Free)
Used as a growth supplement for cell culture media and in other research applications.
Have Questions?
Catalog NumberQuantity
MPB092931949100 mL
Catalog number MPB092931949
Price (SGD)
795.00
EA
Add to cart
Quantity:
100 mL
Price (SGD)
795.00
EA
Add to cart
  • Collected from type AB human blood
  • Used as a growth supplement for cell culture media and in other research applications
  • Proven to grow many human cell lines at a faster rate than mixed blood group serum
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Certifications/ComplianceU.S. regulations
ColorGolden Brown
SpeciesHuman
FormLiquid
Product TypeHuman Serum
Quantity100 mL
SterilitySterile
Sterilization Method0.1 μm filtration
For Use With (Application)Grow many Human cell lines at a faster rate
Hemoglobin Concentration≤26 mg/dL
Mycoplasma DetectedNo
Osmolality263 to 323 mOsm/kg
pH7 to 8
Protein Content5.5 to 7.5 g/dL
Unit SizeEA
Contents & Storage
-20°C