Human Serum: Hepatitis C (HCV)-Free
Human Serum: Hepatitis C (HCV)-Free
Actual product may vary
MP Biomedicals
Human Serum: Hepatitis C (HCV)-Free
Hepatitis Associated Antigen (HBsAG)–free, HIV–free
Have Questions?
Catalog NumberQuantity
MPB092931149100 mL
Catalog number MPB092931149
Price (SGD)
524.00
EA
Add to cart
Quantity:
100 mL
Price (SGD)
524.00
EA
Add to cart
Specifications
Certifications/ComplianceU.S. regulations
ColorGolden Yellow
SpeciesHuman
FormLiquid
Product TypeHuman Serum
Quantity100 mL
SterilitySterile
Sterilization Method0.1 μm filtration
For Use With (Application)Base matrix for hormone and steroid calibrators, controls, standards and linearity products
Hemoglobin Concentration≤26mg/dL
Mycoplasma DetectedNo
Osmolality263 to 323mOsm/kg
pH7 to 8
Protein Content5.5 to 7.5 g/dL
Unit SizeEA
Contents & Storage
-20°C