Serum, Human, Pooled
Serum, Human, Pooled
Actual product may vary
MP Biomedicals
Serum, Human, Pooled
Used as a media component to neutralize cell free virus.
Have Questions?
Catalog NumberQuantity
MPB092930149100 mL
Catalog number MPB092930149
Price (SGD)
452.00
EA
Add to cart
Quantity:
100 mL
Price (SGD)
452.00
EA
Add to cart
Applications: Human pooled serum is used as a media component to neutralize cell free virus. It's used as a reagent in liposome binding assay and in preparation of sperm.
Product Description: This serum is obtained from pooled, normal clotted human blood.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
ColorGolden Brown
FormLiquid
Product TypeHuman Serum
Quantity100 mL
SterilitySterile
For Use With (Application)Media component for cell free virus, liposome binding assay, preparation of sperm
pH7 to 8
Unit SizeEA
Contents & Storage
Store at -20 °C.