Specifications
Certifications/ComplianceUSDA
ColorPale to Golden Brown
FormLiquid
Product TypeFetal Bovine Serum
Quantity1 x 500 mL
Concentration3.1 to 4.4 g/dL
Serum TreatmentHeat-inactivated
SterilitySterile
Endotoxin Concentration<1 ng/mL
For Use With (Application)Used for cell culture and insect cell culture applications
Heat InactivatedYes
Hemoglobin Concentration27 mg/dL
Mycoplasma DetectedNo
Osmolality280 to 325 mOsm/kg
pH6.7 to 7.8
USDA Safety Testing IncludedUSDA Inspected
Unit SizeEach