Phosphate Buffered Saline Tablets (PBS)
Phosphate Buffered Saline Tablets (PBS)
Actual product may vary
Phosphate Buffered Saline Tablets (PBS)
Phosphate Buffered Saline Tablets (PBS)
MP Biomedicals
Phosphate Buffered Saline Tablets (PBS)
Phosphate Buffered Saline Tablets (PBS) are a balanced salt mixture for easy PBS buffer preparation for a variety of applications, including reagent preparation, diluting cells for flow cytometry, and cell culture reagent.
Have Questions?
Catalog NumberQuantity
MPB092810306200 Tablets
Catalog number MPB092810306
Price (SGD)
346.00
EA
Add to cart
Quantity:
200 Tablets
Price (SGD)
346.00
EA
Add to cart
  • No need for weighing and mixing chemicals
  • Simply dissolve PBS tablets in water
  • One tablet dissolved in 100mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
  • Research use only
Specifications
AdditiveNo Calcium, No Magnesium
Product TypePhosphate Buffered Saline (PBS)
Quantity200 Tablets
Molecular FormulaClK
MDL NumberMFCD00011360
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
IUPAC Namepotassium chloride
Molecular Weight (g/mol)74.55
FormTablet
Unit SizeEA