Phosphate Buffered Saline Tablets
Phosphate Buffered Saline Tablets
Actual product may vary
MP Biomedicals
Phosphate Buffered Saline Tablets
Pack of phosphate buffered saline tablets for use in immunoassays or cell culture applications.
Have Questions?
Catalog NumberQuantity
MPB092810305100 Pk.
Catalog number MPB092810305
Price (SGD)
169.00
EA
Add to cart
Quantity:
100 Pk.
Price (SGD)
169.00
EA
Add to cart
Pack of phosphate buffered saline tablets for use in immunoassays or cell culture applications.
Phosphate Buffered Saline Tablets (PBS) are composed of a balanced salt solution with multiple applications in cell culture, cell biology and buffer preparation. This buffer preparation can be used for reagent preparation, diluting cells for flow cytometry and cell culture reagents. One tablet dissolved in 100 mL water yields a buffer containing 0.01 M phosphate, 0.0027 M KCl and 0.137 M NaCl, with a pH of approximately 7.4 at 25°C.
  • Tablet format for convenience and ease of use
  • No need for weighing and mixing chemicals
  • Simply dissolve tablets into water
  • Specifically developed for immunoassay procedures
Specifications
Quantity100 Pk.
Physical FormTablet
Unit SizeEA