Dulbecco's Phosphate Buffered Saline (10X) without Calcium and Magnesium
Actual product may vary
MP Biomedicals
Dulbecco's Phosphate Buffered Saline (10X) without Calcium and Magnesium
Stock solution used to prepare 1X D-PBS in cell culture grade water.
Have Questions?
Catalog NumberQuantity
MPB091960454500 mL
Catalog number MPB091960454
Price (SGD)
78.00
Each
Quantity:
500 mL
Price (SGD)
78.00
Each
Dulbecco's Phosphate Buffered Saline 10X is a stock solution used to prepare 1X D-PBS in cell culture grade water. D-PBS is used to wash cells during preparation and serial transfer. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells. Used to wash cells during preparation and serial transfer.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Chemical Name or MaterialDPBS
Concentration10 X
pH6.5 to 7.5
Physical FormSolution
Quantity500 mL
FormulationLiquid (as 10X) Inorganic Salts: Potassium Chloride [KCl]: 2000.00 mg/L; Potassium Phosphate Monobasic [KH2PO4]: 2000.00 mg/L; Sodium Chloride [NaCl]: 80000.00 mg/L; Sodium Phosphate Dibasic [Na2HPO4]: 11500.00 mg/L
SterilitySterile
IncludesNo Calcium, No Magnesium
GradeNot Available
Unit SizeEach