Check Order Status
1X Phosphate Buffered Saline, Dulbecco's, without Magnesium and Calcium
Actual product may vary
1X Phosphate Buffered Saline, Dulbecco's, without Magnesium and Calcium
Clear, colorless saline solution without magnesium and calcium
Have Questions?
Catalog Number
Quantity
MPB091860454X6
6 x 500 mL
Catalog number
MPB091860454X6
Price (SGD)
462.00
EA
Add to cart
Quantity:
6 x 500 mL
Price (SGD)
462.00
EA
Add to cart
MDL Number: MFCD00131855
Format: Solution
pH: 7 to 7.6
Osmolality: 270 to 305 mM/kg
For handling and culturing of mammalian cells
Specifications
Color
Clear
Dilution
1 X
Product Type
Phosphate Buffered Saline (PBS)
Quantity
6 x 500 mL
Formulation
Potassium Chloride 200mg/L, Potassium Phosphate Monobasic 200mg/L, Sodium Chloride 8000mg/L, Sodium Phosphate Dibasic 1150mg/L
pH
7 to 7.6
Form
Liquid
Unit Size
EA
Contents & Storage
15° to 30°C