Check Order Status
PBS, (1X PBS Liq.), w/o Calcium and Magnesium, 100mL, MP Biomedicals
Actual product may vary
MP Biomedicals
PBS, (1X PBS Liq.), w/o Calcium and Magnesium, 100mL, MP Biomedicals
Dulbecco's Formula Without calcium and magnesium, Storage temperature: 15-30°C
Have Questions?
Catalog Number
Quantity
MPB091860449
100 mL
Catalog number
MPB091860449
Price (SGD)
37.00
Each
Add to cart
Quantity:
100 mL
Price (SGD)
37.00
Each
Add to cart
Specifications
Quantity
100 mL