1X Hanks' Balanced Salt Solution, Modified
Actual product may vary
MP Biomedicals
1X Hanks' Balanced Salt Solution, Modified
Red, Clear Solution
Have Questions?
Catalog NumberQuantity
MPB091810054500 mL
Catalog number MPB091810054
Price (SGD)
63.00
Each
Quantity:
500 mL
Price (SGD)
63.00
Each
Used for washing cells and tissues while maintaining cells in a viable state. Buffered with phosphate and maintains a physiological pH. Contains phenol red.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Chemical Name or MaterialHanks' Balanced Salt Solution (HBSS)
ColorRed
Recommended Storage15°to 30°C
Concentration1 X
pH6.9 to 7.2
Physical FormLiquid
Quantity500 mL
Unit SizeEach