Check Order Status
Trypan Blue Stain Solution, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Trypan Blue Stain Solution, MP Biomedicals
0.4% (w/v) Solution In normal saline Storage temperature 15-30°C
Have Questions?
Catalog Number
Quantity
MPB091691049
100 mL
Catalog number
MPB091691049
Price (SGD)
62.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
62.00
EA
Add to cart
Specifications
Chemical Name or Material
Trypan Blue Stain Solution
CAS
72-57-1
Molecular Formula
C34H24N6Na4O14S4
MDL Number
MFCD00003969
InChI Key
WMYVHJWZUUEZNE-ARWFNKCKSA-J
View more