▸
Buffers and Standards
▸
Life Science Buffers
▸
HEPES Buffer, 1M Solution, Cell Culture Grade, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
HEPES Buffer, 1M Solution, Cell Culture Grade, MP Biomedicals
1M solution pH 7.2 to 7.4 at 37°C Storage temperature: 15-30°C
Have Questions?
Catalog Number
Quantity
MPB091688449
100 mL
Catalog number
MPB091688449
Price (SGD)
98.00
Each
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
98.00
Each
Add to cart
Specifications
Chemical Name or Material
Hepes Buffer
CAS
7365-45-9
Concentration
1 M
Physical Form
Solution
Quantity
100 mL
View more