L-Glutamine, MP Biomedicals
L-Glutamine, MP Biomedicals
L-Glutamine, MP Biomedicals
L-Glutamine, MP Biomedicals
MP Biomedicals
L-Glutamine, MP Biomedicals
200 mM solution Shipped frozen in dry ice
Have Questions?
Catalog NumberQuantity
MPB091680149100 mL
Catalog number MPB091680149
Price (SGD)
62.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
62.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Glutamine
CAS56-85-9
Molecular FormulaC5H10N2O3
MDL NumberMFCD00008044
Synonyml-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
View more