▸
Cell Culture Media, Supplements, and Reagents
▸
Cell Culture Reagents, Antibiotics and Supplements
▸
Antibiotics
▸
Amphotericin B, Antibiotic Solution, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Amphotericin B, Antibiotic Solution, MP Biomedicals
Antibiotic solution Concentration: 250μg/mL Effective against yeast and fungi
Have Questions?
Change view
Quantity:
50 mL
20 mL
Catalog number
MPB091672348
Price (SGD)
131.00
Each
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (SGD)
131.00
Each
Add to cart
Specifications
Chemical Name or Material
Amphotericin B
Type
Amphotericin B
CAS
1397-89-3
Quantity
50 mL
Molecular Formula
C47H73NO17
View more