RPMI 1640 Medium, 1X Liq. without L-Glutamine, L-Cysteine, L-Cystine and L-Methionine
RPMI 1640 Medium, 1X Liq. without L-Glutamine, L-Cysteine, L-Cystine and L-Methionine
Actual product may vary
MP Biomedicals
RPMI 1640 Medium, 1X Liq. without L-Glutamine, L-Cysteine, L-Cystine and L-Methionine
General purpose medium based on the RPMI 1630 line of media
Have Questions?
Catalog NumberQuantity
MPB091646454500 mL
Catalog number MPB091646454
Price (SGD)
149.00
EA
Add to cart
Quantity:
500 mL
Price (SGD)
149.00
EA
Add to cart
RPMI 1640 Medium was originally developed by Moore and his colleagues at Roswell Park Memorial Institute (RPMI). It was based on the RPMI 1630 line of media which utillized a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins.
RPMI has successfully been used for the cultivation of normal human and neoplastic leukocytes. It is now a popular general purpose medium when properly supplemented.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
FormLiquid
Product TypeRPMI 1640
SterilitySterile
Without AdditivesNo Glutamine, No Cystine, No Cysteine, No Methionine
Quantity500 mL
pH6.8 to 7.4
Unit SizeEA
Contents & Storage
4°C