RPMI 1640 Medium, 1X Liq. with L-Glutamine and Sodium Bicarbonate
Actual product may vary
MP Biomedicals
RPMI 1640 Medium, 1X Liq. with L-Glutamine and Sodium Bicarbonate
Used for the cultivation of normal human and neoplastic leukocytes.
Have Questions?
Catalog NumberQuantity
MPB091260354500 mL
Catalog number MPB091260354
Price (SGD)
50.00
Each
Quantity:
500 mL
Price (SGD)
50.00
Each
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
FormLiquid
Product TypeRPMI 1640
SterilitySterile
Quantity500 mL
pH6.9 to 7.4
Unit SizeEach
Contents & Storage
4°C