Alpha Modification of Eagle's Medium (AMEM), Powder
Actual product may vary
MP Biomedicals
Alpha Modification of Eagle's Medium (AMEM), Powder
Alpha Modification of Eagle's Medium (AMEM), powder, with L-glutamine, without deoxyribosides, ribosides, sodium bicarbonate
Have Questions?
Catalog NumberQuantity
MPB09103112010 x 1 L
Catalog number MPB091031120
Price (SGD)
73.00
Each
Quantity:
10 x 1 L
Price (SGD)
73.00
Each
Alpha Modification of Eagle's Medium (AMEM), powder, with L-glutamine, without deoxyribosides, ribosides, sodium bicarbonate. Originally used for growth of hamster kidney cells.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Product TypeMinimum Essential Medium Eagle's
SterilitySterile
With AdditivesGlutamine
Without AdditivesNo Sodium Bicarbonate, No Ribosides, No Deoxyribosides
Quantity10 x 1 L
pH6.5 to 7.5
Unit SizeEach
Contents & Storage
0 to 5°C