Tryptone, Powdered Media
Tryptone, Powdered Media
Actual product may vary
MP Biomedicals
Tryptone, Powdered Media
Pancreatic digest of casein
Have Questions?
Catalog number MPB091010817
Price (SGD)
260.00
EA
Add to cart
Price (SGD)
260.00
EA
Add to cart
Tryptone is a pancreatic digest of casein. It is utillized as a source of nitrogen in many culture media for growing bacteria as well as fungi. Tryptone supplements media for the cultivation and maintenance of fastidious aerobic and facultative microorganisms, such as E. coli and Pseudomonas species. It is used to supplement media in the cultivation of anaerobes.
  • The lack of detectable carbohydrates makes this peptone an excellent choice for bacterial studies based on fermentation reactions.
  • Its high level of tryptophan makes it useful in the production of indol.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
FormPowder
Product TypeMicrobiology Media
For Use With (Application)Cultivation and maintenance of fastidious aerobic and facultative microorganisms such as Escherichia coli and Pseudomonas species
Quantity500 g
Unit SizeEA
Contents & Storage
15 to 30°C