FormPowder
Media TypeTryptic Soy Broth
Product TypeMicrobiology Media
FormulationCasein digest Peptone 17g/L, Soy Peptone 3g/L, Disodium Phosphate 2.5g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L
For Use With (Application)Isolation and cultivation of non-fastidious organisms
Quantity500 g
Unit SizeEach