Transport Factors
Actual product may vary
MP Biomedicals
Transport Factors
Have Questions?
Catalog NumberpH
MPB088234316.8
Catalog number MPB08823431
Price (SGD)
1,593.00
Each
pH:
6.8
Price (SGD)
1,593.00
Each
Transport factors are proteins which enable small metabolites to be carried in usable form to cells. Transferrin binds iron and makes it available to cells in a recognizable form.
Specifications
Product TypeTransferrin
pH6.8
Quantity1 g
SourcePooled Human Plasma
Unit SizeEach
Contents & Storage
-20°C