Product Image
MP Biomedicals
Immuno™ Albumin, Bovine, Low Endotoxin Powder (Heat-Shock)
Used to solubilize lipids, also used as blocking agent in Western blots or ELISA applications.
Have Questions?
Catalog NumberQuantity
MPB08810683100 g
Catalog number MPB08810683
Price (SGD)
714.00
Each
Quantity:
100 g
Price (SGD)
714.00
Each
  • Derived from bovine blood collected at USDA licensed facility
  • Prepared by heat-shocked fractionate method
For Research Use Only.
Specifications
For Use With (Application)ELISA, Immunoassay, Mammalian Cell Culture, Western Blot
Molecular Weight (g/mol)∽66
NameImmuno™ Albumin, Bovine, Low Endotoxin (Heat Shock)
pH Range7.1
Quantity100 g
SourceBovine Blood
Storage Requirements0° to 5°C