▸
Recombinant Proteins
▸
Albumin, Bovine, For IVD, Low IgG
Actual product may vary
MP Biomedicals
Albumin, Bovine, For IVD, Low IgG
Prepared by heat shock fractionation
Have Questions?
Catalog Number
Quantity
MPB08810032
50g
Catalog number
MPB08810032
Price (SGD)
479.00
Each
Add to cart
Quantity:
50g
Price (SGD)
479.00
Each
Add to cart
For IVD applications, immunoassays, enzyme stabilization
Low protease, conductivity and visible absorbance
Purity: >98%
pH: 7
Specifications
For Use With (Application)
Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol)
∽66
Name
Bovine Blood Albumin Fraction V
pH Range
7
Quantity
50g
Source
Bovine Blood
Storage Requirements
0° to 5°C
Form
Crystalline