▸
Isotype Controls and Standards
▸
IgG, Mouse, 10mg
MP Biomedicals
IgG, Mouse, 10mg
Have Questions?
Catalog Number
Quantity
MPB08641461
10 mg
Catalog number
MPB08641461
Price (SGD)
574.00
Each
Add to cart
Quantity:
10 mg
Price (SGD)
574.00
Each
Add to cart
Specifications
Conjugate
Unconjugated
Quantity
10 mg