Check Order Status
Complement, Guinea Pig Purified, 5mL
MP Biomedicals
Complement, Guinea Pig Purified, 5mL
Suitable for use in hemolytic titrations and cytotoxicity assays, and as a serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques
Have Questions?
Catalog Number
Quantity
MPB0855854
5 mL
Catalog number
MPB0855854
Price (SGD)
272.00
EA
Add to cart
Quantity:
5 mL
Price (SGD)
272.00
EA
Add to cart
Specifications
Product Type
Guinea Pig complement
Quantity
5 mL
Unit Size
EA
Contents & Storage
2° to 8°C