▸
Primary Antibodies
▸
All Primary Antibodies
▸
IgG, Whole Molec. PAb, anti-Rabbit, Sheep, Liq. (IgG), HRP, MP Biomedicals
MP Biomedicals
IgG, Whole Molec. PAb, anti-Rabbit, Sheep, Liq. (IgG), HRP, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0855677
2 mL
Catalog number
MPB0855677
Price (SGD)
763.00
Each
Add to cart
Quantity:
2 mL
Price (SGD)
763.00
Each
Add to cart
Specifications
Antigen
IgG, Whole Molecule
Classification
Polyclonal
Conjugate
HRP
Host Species
Sheep
Quantity
2 mL
Primary or Secondary
Primary
Isotype
IgG