Check Order Status
Goat anti-Mouse IgG Fraction, Whole Molecule Polyclonal Secondary Antibody, FITC, MP Biomedicals
MP Biomedicals
Goat anti-Mouse IgG Fraction, Whole Molecule Polyclonal Secondary Antibody, FITC, MP Biomedicals
Have Questions?
Catalog number
MPB0855493
Price (SGD)
395.00
EA
Add to cart
Price (SGD)
395.00
EA
Add to cart
Host Species:
Goat
Isotype:
IgG
Species Reactivity:
Mouse
Specifications
Antigen
IgG, Whole Molecule
Classification
Polyclonal
Conjugate
FITC
Host Species
Goat
Quantity
2 mL
Primary or Secondary
Secondary
Target Species
Mouse
Form
Purified
Isotype
IgG