Goat anti-Human IgM, 5 Fc μ-Specific Polyclonal Secondary Antibody, HRP, MP Biomedicals
Product Image
MP Biomedicals
Goat anti-Human IgM, 5 Fc μ-Specific Polyclonal Secondary Antibody, HRP, MP Biomedicals
Have Questions?
Catalog number MPB0855228
Price (SGD)
671.00
EA
Add to cart
Price (SGD)
671.00
EA
Add to cart
Host Species: Goat
Isotype: IgG
Species Reactivity: Human
Specifications
AntigenIgM, 5 Fc u-Specific
ClassificationPolyclonal
ConjugateHRP
Host SpeciesGoat
Quantity2 mL
Primary or SecondarySecondary
Target SpeciesHuman
FormPurified
IsotypeIgG
Unit SizeEA