Check Order Status
LSM™ Lymphocyte Separation Medium
Actual product may vary
MP Biomedicals
LSM™ Lymphocyte Separation Medium
LSM™ Lymphocyte Separation Medium is a high performance density gradient medium for monocyte separation.
Have Questions?
Catalog Number
Quantity
MPB0850494
5 x 100 mL
Catalog number
MPB0850494
Price (SGD)
510.00
EA
Add to cart
Quantity:
5 x 100 mL
Price (SGD)
510.00
EA
Add to cart
LSM is a sterile-filtered solution of a sucrose polymer and diatrizoate salts
Formulated for isolation of mononuclear cells from defibrinated or heparinized whole human blood
One-step centrifugation permits separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets
Maximum yield of mononuclear cells
> 96% cell viability of lymphocytes
Easy one-step centrifugation
Low endotoxins
Autoclave sterilization
Specifications
Form
Liquid
Product Type
Cell Culture Medium
Sterility
Sterile
Quantity
5 x 100 mL
pH
7.23
Unit Size
EA
Contents & Storage
15° to 30°C