Check Order Status
Zymolyase™
Actual product may vary
MP Biomedicals
Zymolyase™
For lysing the cell walls of viable yeast cells and preparing yeast spheroblasts
Have Questions?
Change view
Catalog Number
Quantity
Concentration
Product Type
MPB08320932
250 mg
100,000 U/mg
Zymolyase 100T
MPB08320931
500 mg
100,000 U/mg
Zymolyase 100T
2 Options
Catalog number
MPB08320932
Price (SGD)
2,515.00
EA
Add to cart
Quantity:
250 mg
Concentration:
100,000 U/mg
Product Type:
Zymolyase 100T
Price (SGD)
2,515.00
EA
Add to cart
Highly purified preparation from
Arthrobacter luteus
Efficiently hydrolyzes poly (β-1,3 glucose)-like yeast cell-wall glucan
One unit is defined as the amount of activity causing a decrease in A
800
of 30%
Available with lytic activity of 20,000U/g (Zymolyase 20T) or 100,000U/g (Zymolyase 100T)
Specifications
Concentration
100,000 U/mg
pH
5 to 10
For Use With (Application)
Used to lyse living yeast cell walls to produce protoplast or spheroplast of various strains of Yeast cells
Source
Arthrobacter luteus
Quantity
250 mg
Product Type
Zymolyase 100T
Form
Lyophilized Powder
Unit Size
EA
Contents & Storage
2°C to 8°C