DNase-, RNase-Free Water, MP Biomedicals™
DNase-, RNase-Free Water, MP Biomedicals™
DNase-, RNase-Free Water, MP Biomedicals™
DNase-, RNase-Free Water, MP Biomedicals™
MP Biomedicals
DNase-, RNase-Free Water, MP Biomedicals™
Suitable for the investigation of nucleic acid and protein synthesis.
Have Questions?
Catalog number MPB04821932
Price (SGD)
112.00
Price (SGD)
112.00
EA
Add to cart
Specifications
CAS7732-18-5
ColorClear, Colorless
Boiling Point100°C
EINECS Number231-791-2
DNaseDNase-free
View more
  • Grade: DNase- RNase-free
  • Key applications: Investigation of nucleic acid and protein synthesis
  • Clear, colorless, liquid
  • 0.2 μ filtered (sterile)