Check Order Status
Water, DNase, RNase Free, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Water, DNase, RNase Free, MP Biomedicals™
Ideal for use with RNase-Erase™ and DNA-Erase™ decontaminants
Have Questions?
Catalog Number
Quantity
MPB04821739
500 mL
Catalog number
MPB04821739
Price (SGD)
158.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
158.00
EA
Add to cart
Specifications
Chemical Name or Material
Water
Name Note
Sterile, Nuclease Free
CAS
7732-18-5
Purity Grade Notes
A sterile solution, completely free of detectable DNase (exo and endo) and RNase; DEPC treated
Molecular Formula
H2O
View more