Guanidine Hydrochloride, Ultra Pure grade, MP Biomedicals
Guanidine Hydrochloride, Ultra Pure grade, MP Biomedicals
Guanidine Hydrochloride, Ultra Pure grade, MP Biomedicals
Guanidine Hydrochloride, Ultra Pure grade, MP Biomedicals
MP Biomedicals
Guanidine Hydrochloride, Ultra Pure grade, MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB048205401 kg
Catalog number MPB04820540
Price (SGD)
702.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
702.00
EA
Add to cart
Specifications
Chemical Name or MaterialGUANIDINE HYDROCHLORIDE
CAS50-01-1
Melting Point180°C to 190°C
ColorWhite
Recommended StorageStore at Room Temperature (15-30°C). Store Desiccated.
View more
  • Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins
  • This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies
  • This can be used as the first step in refolding proteins or enzymes into their active form
  • Urea and dithiothreitol (DTT) may also be necessary
  • Guanidine HCl is used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties
  • It is an inhibitor of RNase
  • Highly concentrated (6 - 8 M) Guanidine HCl solutions are used to denature native globular proteins
  • It apparently disrupts hydrogen bonds which hold the protein in its unique structure; however, there also is evidence suggesting that guanidine hydrocholoride may disrupt hydrophobic interactions by promoting the solubility of hydrophobic residues in aqueous solutions