TRIS, MP Biomedicals™
TRIS, MP Biomedicals™
TRIS, MP Biomedicals™
TRIS, MP Biomedicals™
MP Biomedicals
TRIS, MP Biomedicals™
TRIS Ultra Pure, ≥99.9%, White crystalline powder
Have Questions?
Catalog NumberQuantity
MPB04819620500 g
Catalog number MPB04819620
Price (SGD)
205.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
205.00
EA
Add to cart
Specifications
CAS77-86-1
Melting Point165°C to 175°C
Boiling Point219°C to 220°C (at 10 mmHg)
ColorWhite
Storage Note 1Room Temperature, desiccate
View more
  • Tris is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
  • Tris is used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions.
  • Tris have been useful as buffers in a wide variety of biological systems.
  • It has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).
  • It does not precipitate calcium salts and is of value in maintaining solubility of manganese salts.
  • It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid.
  • It is RNAse and DNAse-free.Tris is relatively non-hygroscopic;but, if needed, it can be dried at 100°C for up to 4 hours to remove any water.