Check Order Status
Sodium Dodecyl Sulfate,Ultra Pure, 100g, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sodium Dodecyl Sulfate,Ultra Pure, 100g, MP Biomedicals
151-21-3 Purity: >>99% (280 nm): <<0.1 Heavy Metals (as Pb): <<0.002% Anionic detergent used to solubilize proteins
Have Questions?
Catalog Number
Quantity
MPB04811034
100 g
Catalog number
MPB04811034
Price (SGD)
145.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
145.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium Dodecyl Sulfate
CAS
151-21-3
Quantity
100 g
Molecular Formula
C12H25NaO4S
MDL Number
MFCD00036175
View more