N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade, MP Biomedicals
N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade, MP Biomedicals
N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade, MP Biomedicals
N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade, MP Biomedicals
MP Biomedicals
N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade, MP Biomedicals
110-18-9 (TEMED) Purity: >99% Polymerization accelerator for polyacrylamide gel formation
Have Questions?
Catalog NumberQuantity
MPB0480561550 mL
Catalog number MPB04805615
Price (SGD)
119.00
EA
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (SGD)
119.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N,N«,N«-Tetramethyl-Ethylenediamine
CAS110-18-9
Molecular FormulaC6H16N2
Quantity50 mL
Synonymtemed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
View more