Check Order Status
Ammonium Persulfate, 100g, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Ammonium Persulfate, 100g, MP Biomedicals
Polymerization catalyst used with TEMED for polyacrylamide gel formation
Have Questions?
Catalog Number
Quantity
MPB04802829
100 g
Catalog number
MPB04802829
Price (SGD)
81.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
81.00
EA
Add to cart
Specifications
Chemical Name or Material
Ammonium Persulfate
CAS
7727-54-0
Quantity
100 g
Molecular Formula
H8N2O8S2
MDL Number
MFCD00003390
View more