Check Order Status
Gelatin Type A; 100 Bloom Food Grade MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Gelatin Type A; 100 Bloom Food Grade MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0296031780
100 g
Catalog number
MPB0296031780
Price (SGD)
194.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
194.00
EA
Add to cart
Specifications
Chemical Name or Material
Gelatin
Name Note
Type A; 100 Bloom
CAS
9000-70-8
MDL Number
MFCD00081638
Quantity
100 g
View more