Choline Deficient Diet With DL-Ethionine, Pellet
Choline Deficient Diet With DL-Ethionine, Pellet
Actual product may vary
MP Biomedicals
Choline Deficient Diet With DL-Ethionine, Pellet
Used in miRNA studies in in vitro and in vivo activated hepatic stellate cells.
Have Questions?
Catalog NumberQuantity
MPB029602141010 kg
Catalog number MPB0296021410
Price (SGD)
5,473.00
EA
Add to cart
Quantity:
10 kg
Price (SGD)
5,473.00
EA
Add to cart
Choline Deficient Diet, Modified: Contains added trace minerals and fibres
Composition (Ingredient, Amount):
  • Vitamin Free Casein, 10%
  • a-Protein, 10%
  • DL-Ethionine, 0.5%
  • Sucrose, 51%
  • Alphacel, Non-Nutritive Bulk, 5%
  • Lard, 20%
  • Manganese Sulfates·H2O, 150 mg/kg diet
  • Zinc Chloride, 20 mg/kg diet
  • Chromium Potassium Sulfate·12H2O, 55 mg/kg diet
  • Sodium Selenite, 1 mg/kg diet
  • Salt Mixture Wesson, 4%
  • Calcium Carbonate, 21% (of total salt mixture)
  • Copper Sulfate·5H20, 0.039% (of total salt mixture)
  • Ferric Phosphate, 1.47% (of total salt mixture)
  • Manganese Sulfate·H2O, 0.02% (of total salt mixture)
  • Magnesium Sulfate·7H2O, 9% (of total salt mixture)
  • Potassium Aluminum Sulfate, 0.009% (of total salt mixture)
  • Potassium Chloride, 12% (of total salt mixture)
  • Potassium Dihydrogen Phosphate, 31% (of total salt mixture)
  • Potassium Iodide, 0.005% (of total salt mixture)
  • Sodium Chloride, 10.5% (of total salt mixture)
  • Sodium Fluoride, 0.057% (of total salt mixture)
  • Tricalcium Phosphate, 14.9% (of total salt mixture)
  • Plus MP Vitamin Diet Fortification Mixture except Choline Chloride
Specifications
Product TypeAnimal Diet
ComponentsVitamin Free Casein 10%, alpha-Soy Protein 10%, DL-Ethionine 0.5%, Sucrose 56%, Lard 20%, AIN Mineral Mix 3.5%, Plus MP Vitamin Diet Fortification Mixture except Choline Chloride
FormatPellet
For Use With (Application)miRNA studies in in-vitro and in-vivo activated hepatic stellate cells
Quantity10 kg
Storage Requirements15°C to 30°C
Unit SizeEA