▸
Biochemical Reagents
▸
Carbohydrates
▸
Glycosyl compounds
▸
Sucrose, ( FCC), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sucrose, ( FCC), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0290471325
25 lb.
Catalog number
MPB0290471325
Price (SGD)
207.00
Each
Add to cart
Quantity:
25 lb.
Request bulk or custom format
Price (SGD)
207.00
Each
Add to cart
Specifications
Chemical Name or Material
Sucrose
CAS
57-50-1
Recommended Storage
Room Temperature
Molecular Formula
C12H22O11
MDL Number
MFCD00006626
View more