Brewers Yeast
Brewers Yeast
Actual product may vary
MP Biomedicals
Brewers Yeast
Used as an insect diet.
Have Questions?
Catalog NumberQuantity
MPB029033122511.33 kg
Catalog number MPB0290331225
Price (SGD)
364.00
EA
Add to cart
Quantity:
11.33 kg
Price (SGD)
364.00
EA
Add to cart
  • Source of B-complex vitamins, chromium and selenium
  • Brewers Yeast is commonly used for baking break, fermenting wine and for beer and ethanol production
Specifications
Product TypeAnimal Diet
FormatPowder
For Use With (Application)Source of B-complex Vitamins, Chromium and Selenium, also used to bake bread, ferment wine, beer and Ethanol production
Quantity11.33 kg
SpeciesS. cerevisiae
Storage Requirements15°C to 30°C
Unit SizeEA