▸
Animal Care and Research
▸
Animal Welfare and Monitoring
▸
Research Animal Foods
▸
Skim Milk Powder
Actual product may vary
MP Biomedicals
Skim Milk Powder
A nonfat dry milk powder is produced from fresh skim milk. Skim Milk Powder is a component used in formulating animal research diets.
Have Questions?
Catalog Number
Quantity
MPB0290288705
5 lb.
Catalog number
MPB0290288705
Price (SGD)
128.00
Each
Add to cart
Quantity:
5 lb.
Price (SGD)
128.00
Each
Add to cart
Skim milk powder
Used in formulating research animal diets
It is heated to achieve denaturation of the whey proteins
Proteins, enzymes & peptides
Specifications
Product Type
Animal Diet
For Use With (Application)
Formulating research animal diet
Quantity
5 lb.
Storage Requirements
Room Temperature