Wesson Salt Mixture
Wesson Salt Mixture
Actual product may vary
MP Biomedicals
Wesson Salt Mixture
Insect diet mixture
Have Questions?
Catalog NumberQuantity
MPB02902851022 kg
Catalog number MPB0290285102
Price (SGD)
181.00
EA
Add to cart
Quantity:
2 kg
Price (SGD)
181.00
EA
Add to cart
Powdered
Wesson's Salt mixture is designed for the insect diet mixture.
Composition (Ingredient, Amount):
  • Calcium Carbonate, 21%
  • Copper Sulfate·5H2O, 0.039%
  • Ferric Phosphate, 1.47%
  • Manganese Sulfate·H2O, 0.02%
  • Magnesium Sulfate·7H2O, 9%
  • Potassium Aluminum Sulfate, 0.009%
  • Potassium Chloride, 12%
  • Potassium Phosphate Monobasic, 31%
  • Potassium Iodide, 0.005%
  • Sodium Chloride, 10.5%
  • Sodium Fluoride, 0.057%
  • Tricalcium Phosphate, 14.9%
Preparation:
  • Make 1 X solution by dissolving 8 g of powder in 1 L of distilled water
Specifications
Product TypeAnimal Diet
ComponentsCalcium Carbonate 21%, Copper Sulfate.5H2O 0.039%, Ferric Phosphate 1.47%, Man ganese Sulfate.H2O 0.02%, Ma gnesium Sulfate.7H2O 9%, Potassium Aluminum Sulfate 0.009%, Potassium Chloride 12%, Potassium Phosphate Monobasic 31%, Potassium Iodide 0.005%, Sodium Chloride 10.5%, Sodium Fluoride 0.057%, Tricalcium Phosphate 14.9%
FormatPowder
Quantity2 kg
SpeciesRat
Storage Requirements15°C to 30°C
Unit SizeEA