Liver Powder
Actual product may vary
MP Biomedicals
Liver Powder
From Bovine Liver
Have Questions?
Catalog NumberQuantity
MPB0290039601450 g
Catalog number MPB0290039601
Price (SGD)
109.00
Each
Quantity:
450 g
Price (SGD)
109.00
Each
A natural food for increased strength and endurance, beef liver is full of vitamin B-12, iron, and a host of other beneficial nutrients. It often assists in repairing damaged nerve tissue.
Specifications
Product TypeAnimal Diet
FormatLyophilized
For Use With (Application)Liver powder was used to study reactive Oxygen species production and Brugia pahangi Survivorship in Aedes polynesiensis with Artificial Wolbachia Infection Types
Quantity450 g
Storage Requirements15°C to 30°C
Unit SizeEach