Check Order Status
Paraformaldehyde Solution, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Paraformaldehyde Solution, MP Biomedicals™
Paraformaldehyde Solution, Methanol Free, clear Liquid
Have Questions?
Change view
Quantity:
10 x 10 mL
2 x 10 mL
Catalog number
MPB0219998380
Price (SGD)
559.00
EA
Add to cart
Quantity:
10 x 10 mL
Request bulk or custom format
Price (SGD)
559.00
EA
Add to cart
Specifications
CAS
30525-89-4
Storage Note 1
+4°C, store under nitrogen
Molecular Formula
CH2O
Quantity
10 x 10 mL
InChI Key
WSFSSNUMVMOOMR-UHFFFAOYSA-N
View more
Recommended for use in the preparation of formalin fixatives for tissues or cells when the samples are to be used in florescence studies.
Methanol free Paraformaldehyde is ready to use solution used as a fixative for electron microscopy.