CellMaxx™ Bovine Albumin, Low Free Fatty Acid, Stem Cell Culture
Actual product may vary
MP Biomedicals
CellMaxx™ Bovine Albumin, Low Free Fatty Acid, Stem Cell Culture
Bovine serum albumin suitable for stem cell culture and media formulation requiring very low levels of lipids
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
MPB02199899911000 g
MPB0219989980100 g
MPB0219989990500 g
Catalog number MPB0219989991
Price (SGD)
5,547.00
Each
Quantity:
1000 g
Price (SGD)
5,547.00
Each
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Features low endotoxin, low level of lipids, fatty acids and hormones, e.g. insulin, T3, T4.
Physical characteristics:
  • Form: Off white to brown powder
  • Purity: ≥98% (w/w)
  • Total Protein: ≥96%
  • Free Fatty Acids ≤0.005% w/w
  • pH (4%): 6.5 to 7.5
  • Solubility: Dissolves in 20 minutes at 20° to 25°C
  • Moisture: ≤5% w/w
Microbial evaluation and control:
  • Endotoxin: ≤1EU/mg
  • Mycoplasma: Negative
  • Bioburden: ≤100cfu/g
  • Virus (9CFR): Negative
  • Reported: Insulin, T3, T4
Certifications: ISO9000, cGMP, EDQM
Specifications
For Use With (Application)Mammalian Cell Culture
Molecular Weight (g/mol)66
NameCellMaxx™ Bovine Albumin
pH Range6.5 to 7.5
Purification MethodChromatography
Quantity1000 g
SourceBovine
Storage Requirements4°C