Kanamycin sulfate from Streptomyces kanamycetikus, MP Biomedicals™
Kanamycin sulfate from Streptomyces kanamycetikus USP Grade, White Powder
Have Questions?
Catalog Number
Quantity
MPB0219985305
5 g
Catalog number MPB0219985305
Price (SGD)
292.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
292.00
EA
Add to cart
Specifications
Physical FormPowder
CAS25389-94-0
pH6.5 to 8.5 (1% solution)
Melting Point>250°C
Storage Note 1Store at Room Temperature (15-30°C).
View more
It is a broad spectrum aminoglycoside-antibiotic derived from Streptomyces kanamyceticus.
It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium.
Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene.
Kanamycin sulfate is an antimicrobial agent effective against Gram-negative and Gram-positive bacteria and mycoplasma.
It binds to the 70S ribosomal subunit, inhibits translocation, and elicits miscoding.
Cross-resistance occurs between kanamycin and neomycin, framycetin, and paromomycin, and partial cross-resistance has been reported between kanamycin and streptomycin. Aminoglycosidemodifying enzymes (acetyltransferase, phosphotransferase, nucleotidyltransferase) alter the antibiotic, preventing its interaction with ribosomes.