Sodium Phosphate Monobasic, Monohydrate, MP Biomedicals™
Sodium Phosphate Monobasic, Monohydrate, MP Biomedicals™
Sodium Phosphate Monobasic, Monohydrate, MP Biomedicals™
Sodium Phosphate Monobasic, Monohydrate, MP Biomedicals™
MP Biomedicals
Sodium Phosphate Monobasic, Monohydrate, MP Biomedicals™
Features a very high buffering capacity. Sodium Phosphate Monobasic, Monohydrate, MP Biomedicals is a reagent with a variety of laboratory, industrial and manufacturing uses. Most neutral sodium phosphate buffer solutions consist of mixtures of Sodium Pho
Have Questions?
Catalog NumberQuantity
MPB02199202.5500 g
Catalog number MPB02199202.5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
CAS10049-21-5
Recommended StorageStore at Room Temperature (15–30°C), desiccate.
Physical FormPowder
Quantity500 g
Assay Percent Range98.0–103.0%
View more
Sodium phosphate occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4). Most neutral sodium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.
  • A table for preparation of 0.1M sodium phosphate buffer at 25°C using various proportions of sodium phosphate monobasic and sodium phosphate dibasic has been published.
  • A study of the effect of freeze-thaw storage cycles on proteins in sodium phosphate and potassium phosphate buffer solutions has been reported.
  • The effect of 5mM sodium phosphate on the efficacy of electrospray ionization (ESI) ion mobility spectrometry (IMS) analysis has been evaluated.
  • A protocol for the purification of pyrogen-free mouse IgG1 monoclonal antibodies which uses 10 mM sodium phosphate (pH 7.4) has been published.
  • An ion-pairing HPLC method for the analysis of 5-aminosalicylic acid has been reported.
  • A TLC method for separation of nucleotide sugars in the study of glycosyltransferase activity has been published.
Sodium phosphate monobasic, monohydrate has a wide variety of uses:
  • Sequestrant, emulsifier and buffer in foods
  • Mordant in dyeing
  • Weighting silk in tanning
  • Manufacture of enamels, ceramics, detergents, boiler compounds
  • Fireproofing agent
  • In soldering and brazing instead of borax
  • Reagent and buffer in analytical chemistry
  • Cathartic
  • Laxative