N-Formylmethionine Specific Transfer Ribonucleic From E. coli MRE 600, MP Biomedicals™
N-Formylmethionine Specific Transfer Ribonucleic From E. coli MRE 600 Activity=1300 ± 50 picomoles per A260 units, White Powder
Have Questions?
Change view
Quantity:
50 U
10 U
Catalog number MPB0219915450
Price (SGD)
-
Quantity:
50 U
Request bulk or custom format
Specifications
CAS63231-63-0
Storage Note 1-20°C
ColorWhite
MDL NumberMFCD00132196
Quantity50 U
View more
Ribonucleic acid, transfer, N-formylmethionine-specific may be used as a substrate to study the specifity and kinetics of natural, mutated and chemically engineered N-formylmethionine-specific E. coli aminoacyl-tRNA synthetase(s) and methionyl-tRNA formyltransferase(s) (MTF).
Each T-RNA molecule is specific for and binds with a particular amino acid; more than one exists for each amino acid.
T-RNA performs three functions during protein synthesis: binds with its specific amino acid, recognizes the corresponding codon on mRNA and places the amino acid in the correct position for attachment to the polypeptide chain being formed & binds the polypeptide to the ribosome.